Thymol blue sodium salt ACS
Supplier: Thermo Fisher Scientific
Synonyms:
Sodium hydrogen 4,4'-(3H-2,1-benzoxathiol-3-ylidene)bis[5-methyl-2-(1-methylethyl)phenolate] S,S-dioxide, Sodium 4-(3-(4-hydroxy-5-isopropyl-2-methylphenyl)-1,1-dioxido-3H-benzo[c][1,2]oxathiol-3-yl)-2-isopropyl-5-methylphenolate, Thymol blue water soluble, Thymol Blue sodium
42785.04
42785.09
42785.04EA
0
EUR
42785.04
42785.09
Thymol blue sodium salt ACS
Thymol blue sodium salt
CAS No.: 62625-21-2
Molecular Formula: C27H29NaO5S
Formula Weight: 488.58
Physical Form: Crystalline
Melting Point: ca 284° dec.
Solubility: Soluble in water
Beilstein Reference: 3861515
MDL No.: MFCD00151093
Water soluble pH indicator
Formula:
C₂₇H₂₉NaO₅S MW: 488.58 g/mol Melting Pt: ∼ 284 °C (dec.) Density: 1.19 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00151093 CAS Number: 62625-21-2 EINECS: 263-650-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...