Anti-CD8a Mouse Monoclonal Antibody [clone: SK1] (PE)

Supplier: AAT BIOQUEST
100811L0 100811L1 100811L2
AATB100811L0EA 271 EUR
AATB100811L0 AATB100811L1 AATB100811L2
Anti-CD8a Mouse Monoclonal Antibody [clone: SK1] (PE)
Antibodies
SK1 is an anti-human monoclonal antibody that is specific for the CD8a antigen.

  • Particularly useful for flow cytometry

This antibody was purified through affinity chromatography.

Caution: For research use only.

Type: Primary
Antigen: CD8a
Clonality: Monoclonal
Clone: SK1
Conjugation: PE
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR