Anti-CD19 Mouse Monoclonal Antibody [clone: HIB19] (iFluor® 350)

Supplier: AAT BIOQUEST
10192010 10192011
AATB10192010EA 371 EUR
AATB10192010 AATB10192011
Anti-CD19 Mouse Monoclonal Antibody [clone: HIB19] (iFluor® 350)
Antibodies
HIB19 is an anti-human monoclonal antibody that recognises the CD19 antigen.

  • Particularly useful for flow cytometry

This antibody was purified through affinity chromatography.

Caution: For research use only.

Type: Primary
Antigen: CD19 antigen
Clonality: Monoclonal
Clone: HIB19
Conjugation: iFluor® 350
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR