Anti-CD44 Rat Monoclonal Antibody [clone: IM7] (iFluor® 820)

Supplier: AAT BIOQUEST
104410P0 104410P1
AATB104410P0EA 371 EUR
AATB104410P0 AATB104410P1
Anti-CD44 Rat Monoclonal Antibody [clone: IM7] (iFluor® 820)
Antibodies
IM7 is an anti-human/mouse monoclonal antibody that targets the CD44 antigen.

  • Particularly useful for flow cytometry

This antibody was purified through affinity chromatography.

Caution: For research use only.

Type: Primary
Antigen: CD44 antigen
Clonality: Monoclonal
Clone: IM7
Conjugation: iFluor® 820
Epitope:
Host: Rat
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR