Anti-CD14 Mouse Monoclonal Antibody [clone: 61D3] (PerCP/Cy5.5)

Supplier: AAT BIOQUEST
101411U0 101411U1 101411U2
AATB101411U0EA 319 EUR
AATB101411U0 AATB101411U1 AATB101411U2
Anti-CD14 Mouse Monoclonal Antibody [clone: 61D3] (PerCP/Cy5.5)
Antibodies
61D3 is an anti-human monoclonal antibody that targets the CD14 antigen.

  • Particularly useful for flow cytometry

This antibody was purified through affinity chromatography.

Caution: For research use only.

Type: Primary
Antigen: CD14 antigen
Clonality: Monoclonal
Clone: 61D3
Conjugation: PerCP/Cy5.5
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR