Anti-CD14 Mouse Monoclonal Antibody [clone: 61D3] (mFluor™ UV375)

Supplier: AAT BIOQUEST
101410X0 101410X1
AATB101410X0EA 371 EUR
AATB101410X0 AATB101410X1
Anti-CD14 Mouse Monoclonal Antibody [clone: 61D3] (mFluor™ UV375)
Antibodies
61D3 is an anti-human monoclonal antibody that targets the CD14 antigen.

  • Particularly useful for flow cytometry

This antibody was purified through affinity chromatography.

Caution: For research use only.

Type: Primary
Antigen: CD14
Clonality: Monoclonal
Clone: 61D3
Conjugation: mFluor™ UV375
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR