Anti-CD28 Mouse Monoclonal Antibody [clone: CD28.2] (PB (Pacific Blue))

Supplier: AAT BIOQUEST
102801J0 102801J1
AATB102801J0EA 371 EUR
AATB102801J0 AATB102801J1
Anti-CD28 Mouse Monoclonal Antibody [clone: CD28.2] (PB (Pacific Blue))
Antibodies
CD28.2 is an anti-human monoclonal antibody that forms an immune complex with the CD28 antigen.

  • Particularly useful for flow cytometry

This antibody was purified through affinity chromatography.

Caution: For research use only.

Type: Primary
Antigen: CD28
Clonality: Monoclonal
Clone: CD28.2
Conjugation: PB (Pacific Blue)
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR