Anti-CD28 Mouse Monoclonal Antibody [clone: CD28.2] (PerCP (Peridinin-Chlorophyll Protein Complex))

Supplier: AAT Bioquest
102801T0 102801T1 102801T2
AATB102801T0EA 271 EUR
AATB102801T0 AATB102801T1 AATB102801T2
Anti-CD28 Mouse Monoclonal Antibody [clone: CD28.2] (PerCP (Peridinin-Chlorophyll Protein Complex))
Antibodies
CD28.2 is an anti-human monoclonal antibody that forms an immune complex with the CD28 antigen.

  • Particularly useful for flow cytometry

This antibody was purified through affinity chromatography.

Caution: For research use only.

Type: Primary
Antigen: CD28
Clonality: Monoclonal
Clone: CD28.2
Conjugation: PerCP (Peridinin-Chlorophyll Protein Complex)
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR