Wood's alloy (lipowitz's alloy)
Supplier: Roth Carl
Synonyms:
Wood's metal, Bismuth Lead Cadmium Tin alloy (50:25:12.5:12.5 w%)
1980.1
1980.2
ROTH1980.1EA
0
EUR
ROTH1980.1
ROTH1980.2
Wood's alloy (lipowitz's alloy)
Wood's alloy (Lipowitz's alloy)
Melting Pt:
75 °C Density: 9,7 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00198175 CAS Number: 76093-98-6 |