Sulphurous acid
Supplier: Roth Carl
Synonyms:
Sulphur dioxide solution
9328.1
9328.2
ROTH9328.1EA
42.5
EUR
ROTH9328.1
ROTH9328.2
Sulphurous acid
Sulfurous acid
Formula:
H₂O₃S MW: 82,08 g/mol Density: 1,03 g/cm³ (20 °C) |
MDL Number:
MFCD00011335 CAS Number: 7782-99-2 EINECS: 231-973-1 UN: 1833 ADR: 8,II Merck Index: 12,09149 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...