Anti-CD8a Rat Monoclonal Antibody [clone: 53-6.7] (PE)

Supplier: STEMCELL Technologies
60023PE 60023PE.1
STMC60023PEEA 308 EUR
STMC60023PE STMC60023PE.1
Anti-CD8a Rat Monoclonal Antibody [clone: 53-6.7] (PE)
Antibodies
Rat monoclonal IgG2a antibody against mouse, toad CD8a, PE-conjugated.

  • T cells
  • T cells CD8

The 53-6.7 antibody reacts with murine CD8a, a 32 to 34 kDa type I transmembrane glycoprotein which is a subunit of CD8. CD8 is a disulfide-bonded dimer, found either as a heterodimer of CD8a (α) and CD8b (β) subunits (i.e., αβ) or a homodimer (αα). CD8 acts as a co-receptor to the T cell receptor (TCR) during T cell activation by binding MHC Class I molecules presented by an antigen-presenting cell. It functions to strengthen the association between the TCR andMore Product Information
Order Now

SPECIFICATIONS

Antigen name CD8a
Environmentally Preferable
Clonality Monoclonal
Conjugation PE (Phycoerythrin)
Clone 53-6.7
Reactivity Frog,Mouse
Host Rat
Gene ID 12525
Isotype IgG2a kappa

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR