Anti-PD-L2 Rabbit Recombinant Antibody [clone: DM169]

Supplier: ANTIBODIES.COM
A318540-100 A318540-500
ANTIA318540-100EA 640 EUR
ANTIA318540-100 ANTIA318540-500
Anti-PD-L2 Rabbit Recombinant Antibody [clone: DM169]
Antibodies
Recombinant rabbit monoclonal [DM169] antibody to PD-L2 for ELISA and Flow cytometry with samples derived from human.

  • Validated applications: ELISA, Flow cytometry
  • Recommended dilutions: ELISA: 1:5000 to 10000, Flow cytometry: 1:100

Type: Primary
Antigen: PD-L2
Clonality: Recombinant
Clone: DM169
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR