Anti-IgA Mouse Monoclonal Antibody [clone: KT13]

Supplier: ANTIBODIES.COM
A17-100 A17-200
ANTIA17-100EA 445 EUR
ANTIA17-100 ANTIA17-200
Anti-IgA Mouse Monoclonal Antibody [clone: KT13]
Antibodies
Mouse monoclonal (KT13) antibody to IgA for ELISA and WB with samples derived from human.

  • Validated applications: ELISA, WB
  • Recommended dilutions: ELISA: 1 µg/ml, WB: 1/1000 to 1/20000

Type: Primary
Antigen: IgA
Clonality: Monoclonal
Clone: KT13
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR