Anti-IgG Antibody

Supplier: ANTIBODIES.COM
A188-100 A188-200
ANTIA188-100EA 445 EUR
ANTIA188-100 ANTIA188-200
Anti-IgG Antibody
Antibodies
  • Validated applications: ELISA, WB
  • Recommended dilutions: ELISA: 1/1000 to 1/5000, WB: 1/500 to 1/10000

Type: Primary
Antigen: IgG
Clonality:
Clone:
Conjugation: Unconjugated
Epitope:
Host:
Isotype:
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR