Anti-IL-22 Recombinant Recombinant Antibody [clone: DMC471]

Supplier: Antibodies.com
A318719-100 A318719-500
ANTIA318719-100EA 640 EUR
ANTIA318719-100 ANTIA318719-500
Anti-IL-22 Recombinant Recombinant Antibody [clone: DMC471]
Antibodies
Recombinant chimeric monoclonal [DMC471] antibody to IL-22 for flow cytometry with samples derived from human.

  • Validated applications: Flow cytometry
  • Recommended dilutions: Flow cytometry: 1:100

Type: Primary
Antigen: IL-22
Clonality: Recombinant
Clone: DMC471
Conjugation: Unconjugated
Epitope:
Host: Recombinant
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR