Anti-mCherry Goat Polyclonal Antibody

Supplier: Antibodies.com
A270547-100 A270547-50
ANTIA270547-100EA 520 EUR
ANTIA270547-100 ANTIA270547-50
Anti-mCherry Goat Polyclonal Antibody
Antibodies
Goat polyclonal antibody to mCherry for WB, ICC/IF and IHC.

  • Validated applications: WB, ICC/IF, IHC
  • Recommended dilutions: WB: 1:1000, ICC/IF: 1:500, IHC: 1:500

Type: Primary
Antigen: mCherry
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR