Anti-CD86 Rabbit Recombinant Antibody [clone: DM86] (Biotinylated)

Supplier: ANTIBODIES.COM
A317371-100 A317371-500
ANTIA317371-100EA 925 EUR
ANTIA317371-100 ANTIA317371-500
Anti-CD86 Rabbit Recombinant Antibody [clone: DM86] (Biotinylated)
Antibodies
Recombinant rabbit monoclonal [DM86] antibody to CD86 (Biotin) for ELISA and Flow Cytometry with samples derived from Human.

  • Validated applications: ELISA, Flow cytometry
  • Recommended dilutions: ELISA: 1:5000 to 10000, Flow cytometry: 1:100

Type: Primary
Antigen: CD86
Clonality: Recombinant
Clone: DM86
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR