Anti-CD33 Rabbit Recombinant Antibody [clone: DM77] (Biotinylated)

Supplier: ANTIBODIES.COM
A317383-100 A317383-500
ANTIA317383-100EA 925 EUR
ANTIA317383-100 ANTIA317383-500
Anti-CD33 Rabbit Recombinant Antibody [clone: DM77] (Biotinylated)
Antibodies
Recombinant rabbit monoclonal [DM77] antibody to CD33 (Biotin) for ELISA and Flow Cytometry with samples derived from Human.

  • Validated applications: ELISA, Flow cytometry
  • Recommended dilutions: ELISA: 1:5000 to 10000, Flow cytometry: 1:100

Type: Primary
Antigen: CD33
Clonality: Recombinant
Clone: DM77
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR