Anti-CD80 Rabbit Recombinant Antibody [clone: DM110]

Supplier: ANTIBODIES.COM
A318597-100 A318597-500
ANTIA318597-100EA 640 EUR
ANTIA318597-100 ANTIA318597-500
Anti-CD80 Rabbit Recombinant Antibody [clone: DM110]
Antibodies
Recombinant rabbit monoclonal [DM110] antibody to CD80 for ELISA and flow cytometry with samples derived from human.

  • Validated applications: ELISA, Flow cytometry
  • Recommended dilutions: ELISA: 1:5000 to 10000, Flow cytometry: 1:100

Type: Primary
Antigen: CD80
Clonality: Recombinant
Clone: DM110
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR