Anti-Synaptophysin Mouse Monoclonal Antibody [clone: ABT-SYN]

Supplier: ANTIBODIES.COM
A97802-100 A97802-50
ANTIA97802-100EA 475 EUR
ANTIA97802-100 ANTIA97802-50
Anti-Synaptophysin Mouse Monoclonal Antibody [clone: ABT-SYN]
Antibodies
Mouse monoclonal (ABT-SYN) antibody to Synaptophysin for IHC with samples derived from human.

  • Validated applications: IHC

Type: Primary
Antigen: Synaptophysin
Clonality: Monoclonal
Clone: ABT-SYN
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR