Anti-Mucin 5AC Mouse Monoclonal Antibody [clone: ABT-MUC5AC]

Supplier: ANTIBODIES.COM
A98940-100 A98940-50
ANTIA98940-100EA 475 EUR
ANTIA98940-100 ANTIA98940-50
Anti-Mucin 5AC Mouse Monoclonal Antibody [clone: ABT-MUC5AC]
Antibodies
Mouse monoclonal (ABT-MUC5AC) antibody to MUC5AC for IHC with samples derived from human.

  • Validated applications: IHC

Type: Primary
Antigen: Mucin 5AC
Clonality: Monoclonal
Clone: ABT-MUC5AC
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR