Anti-p27 KIP 1 Mouse Monoclonal Antibody [clone: ABT-p27]

Supplier: ANTIBODIES.COM
A99498-100 A99498-50
ANTIA99498-100EA 475 EUR
ANTIA99498-100 ANTIA99498-50
Anti-p27 KIP 1 Mouse Monoclonal Antibody [clone: ABT-p27]
Antibodies
Mouse monoclonal (ABT-p27) antibody to p27Kip1 for IHC with samples derived from human.

  • Validated applications: IHC

Type: Primary
Antigen: p27 KIP 1
Clonality: Monoclonal
Clone: ABT-p27
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR