Anti-Sall4 Mouse Monoclonal Antibody [clone: ABT-SALL4]

Supplier: ANTIBODIES.COM
A99685-100 A99685-50
ANTIA99685-100EA 475 EUR
ANTIA99685-100 ANTIA99685-50
Anti-Sall4 Mouse Monoclonal Antibody [clone: ABT-SALL4]
Antibodies
Mouse monoclonal (ABT-SALL4) antibody to SALL4 for IHC with samples derived from human.

  • Validated applications: IHC

Type: Primary
Antigen: Sall4
Clonality: Monoclonal
Clone: ABT-SALL4
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR