Anti-Hsp60 Mouse Monoclonal Antibody [clone: ABT-HSP60]

Supplier: ANTIBODIES.COM
A99687-100 A99687-50
ANTIA99687-100EA 475 EUR
ANTIA99687-100 ANTIA99687-50
Anti-Hsp60 Mouse Monoclonal Antibody [clone: ABT-HSP60]
Antibodies
Mouse monoclonal (ABT-HSP60) antibody to HSP60 for IHC with samples derived from human.

  • Validated applications: IHC

Type: Primary
Antigen: Hsp60
Clonality: Monoclonal
Clone: ABT-HSP60
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR