Anti-GFP Rabbit Polyclonal Antibody

Supplier: ANTIBODIES.COM
A85298-100 A85298-50
ANTIA85298-100EA 520 EUR
ANTIA85298-100 ANTIA85298-50
Anti-GFP Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody to GFP for WB and ICC/IF.

  • Validated applications: WB, ICC/IF
  • Recommended dilutions: WB: 1:1000 to 1:5000, ICC/IF: 1:2000 to 1:5000
  • Host: Rabbit
  • Clonality: Polyclonal
  • Isotype: IgG
  • Conjugate: Unconjugat

Type: Primary
Antigen: GFP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR