Anti-PI 3 Kinase catalytic subunit alpha Mouse Monoclonal Antibody [clone: ABT-PI3.H1047R]

Supplier: ANTIBODIES.COM
A95056-100 A95056-50
ANTIA95056-100EA 475 EUR
ANTIA95056-100 ANTIA95056-50
Anti-PI 3 Kinase catalytic subunit alpha Mouse Monoclonal Antibody [clone: ABT-PI3.H1047R]
Antibodies
Mouse monoclonal (ABT-PI3.H1047R) antibody to PIK3CA H1047R for IHC and WB with samples derived from Human.

  • Validated Applications: IHC, WB

Type: Primary
Antigen: PI 3 Kinase catalytic subunit alpha
Clonality: Monoclonal
Clone: ABT-PI3.H1047R
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR