Anti-smooth muscle Actin Mouse Monoclonal Antibody [clone: ABT-SMA]

Supplier: ANTIBODIES.COM
A95677-100 A95677-50
ANTIA95677-100EA 475 EUR
ANTIA95677-100 ANTIA95677-50
Anti-smooth muscle Actin Mouse Monoclonal Antibody [clone: ABT-SMA]
Antibodies
Mouse monoclonal (ABT-SMA) antibody to smooth muscle Actin for IHC with samples derived from Human.

  • Validated Applications: IHC

Type: Primary
Antigen:
Clonality: Monoclonal
Clone: ABT-SMA
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR