Anti-Cytokeratin 6a Mouse Monoclonal Antibody [clone: ABT-CK6]

Supplier: Antibodies.com
A96297-100 A96297-50
ANTIA96297-100EA 475 EUR
ANTIA96297-100 ANTIA96297-50
Anti-Cytokeratin 6a Mouse Monoclonal Antibody [clone: ABT-CK6]
Antibodies
Mouse monoclonal (ABT-CK6) antibody to Cytokeratin 6 for IHC and WB with samples derived from Human.

  • Validated Applications: IHC, WB

Type: Primary
Antigen: Cytokeratin 6a
Clonality: Monoclonal
Clone: ABT-CK6
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR