Anti-Cytokeratin 20 Mouse Monoclonal Antibody [clone: ABT-Ck20]

Supplier: Antibodies.com
A96671-100 A96671-50
ANTIA96671-100EA 475 EUR
ANTIA96671-100 ANTIA96671-50
Anti-Cytokeratin 20 Mouse Monoclonal Antibody [clone: ABT-Ck20]
Antibodies
Mouse monoclonal (ABT-Ck20) antibody to Cytokeratin 20 for IHC with samples derived from Human.

  • Validated Applications: IHC

Type: Primary
Antigen: Cytokeratin 20
Clonality: Monoclonal
Clone: ABT-Ck20
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR