Anti-Chromogranin A Mouse Monoclonal Antibody [clone: ABT-CGA]

Supplier: ANTIBODIES.COM
A97063-100 A97063-50
ANTIA97063-100EA 475 EUR
ANTIA97063-100 ANTIA97063-50
Anti-Chromogranin A Mouse Monoclonal Antibody [clone: ABT-CGA]
Antibodies
Mouse monoclonal (ABT-CGA) antibody to Chromogranin A for IHC with samples derived from Human.

  • Validated Applications: IHC

Type: Primary
Antigen: Chromogranin A
Clonality: Monoclonal
Clone: ABT-CGA
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR