Anti-CD43 Mouse Monoclonal Antibody [clone: ABT-CD43]

Supplier: ANTIBODIES.COM
A100029-100 A100029-50
ANTIA100029-100EA 475 EUR
ANTIA100029-100 ANTIA100029-50
Anti-CD43 Mouse Monoclonal Antibody [clone: ABT-CD43]
Antibodies
Mouse monoclonal (ABT-CD43) antibody to CD43 for IHC with samples derived from human.

  • Validated Applications: IHC

Type: Primary
Antigen: CD43
Clonality: Monoclonal
Clone: ABT-CD43
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR