Anti-CD23 Mouse Monoclonal Antibody [clone: ABT-CD23]

Supplier: ANTIBODIES.COM
A100033-100 A100033-50
ANTIA100033-100EA 475 EUR
ANTIA100033-100 ANTIA100033-50
Anti-CD23 Mouse Monoclonal Antibody [clone: ABT-CD23]
Antibodies
Mouse monoclonal (ABT-CD23) antibody to CD23 for IHC with samples derived from human.

  • Validated Applications: IHC

Type: Primary
Antigen: CD23
Clonality: Monoclonal
Clone: ABT-CD23
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR