Anti-TSH alpha Mouse Monoclonal Antibody [clone: ABT-HCG]

Supplier: ANTIBODIES.COM
A100701-100 A100701-50
ANTIA100701-100EA 475 EUR
ANTIA100701-100 ANTIA100701-50
Anti-TSH alpha Mouse Monoclonal Antibody [clone: ABT-HCG]
Antibodies
Mouse monoclonal (ABT-HCG) antibody to hCG for IHC with samples derived from human.

  • Validated Applications: IHC

Type: Primary
Antigen: TSH alpha
Clonality: Monoclonal
Clone: ABT-HCG
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR