Portable Vaccine Coolers
Supplier: GLEN DIMPLEX
VCP18UK VCP8UK
GLNDVCP18UKEA 270 EUR
GLNDVCP18UK GLNDVCP8UK
Portable Vaccine Coolers
Coolers Transport Coolers
Portable vaccine cooler with foil liner and reusable gel packs.
  • 2 to 8 °C operating temperature range
  • External LED temperature display
  • Holds temperature inside between 2 and 8 °C for up to 30 hours
  • Aluminium heat preserving foil lining
  • Covered bung non slip feet
  • Webbing adjustable shoulder strap
  • Comes with 9 and 12 gel packs