3-Amino-2-naphthoic acid 97%
Supplier: Thermo Fisher Scientific
|
Warning
|
Synonyms:
3-Amino-2-naphthoic Acid
L19312.03
L19312.06
L19312.03EA
45.8
EUR
L19312.03
L19312.06
3-Amino-2-naphthoic acid 97%
3-Amino-2-naphthoic acid
Purity: 97%
CAS No.: 5959-52-4
Molecular Formula: C11H9NO2
Formula Weight: 187.20
Melting Point: ca 210° dec.
Beilstein Reference: 744099
Merck Reference: 14,452
MDL No.: MFCD00004115
Formula:
C₁₁H₉NO₂ MW: 187.2 g/mol Boiling Pt: 383 Melting Pt: ∼ 210 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00004115 CAS Number: 5959-52-4 EINECS: 227-726-2 Merck Index: 13,00451 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...