Diisopropylcarbodiimide, Sigma-Aldrich®
Supplier: Merck
|
Danger
|
Synonyms:
N,N'-methanetetraylbis(1-methylethylamine), N,N-Diisopropylcarbodiimide, 1,3-Diisopropylcarbodiimide, N,N'-Diisopropylcarbodiimide, N.N’-Diisopropylcarbodiimide, DIC
M-Clarity™ quality level = MQ200
8.03649.0010
8.03649.0025
8.03649.0100
8.03649.1000
8.03649.0010EA
57.8
EUR
8.03649.0010
8.03649.0025
8.03649.0100
8.03649.1000
Diisopropylcarbodiimide, Sigma-Aldrich®
Diisopropylcarbodiimide
Formula:
C₇H₁₄N₂ Boiling Pt: 145 °C (1013 hPa) Density: 0,81 g/cm³ (20 °C) Flash Pt: 33 °C Storage Temperature: Ambient |
MDL Number:
MFCD00065689 CAS Number: 693-13-0 EINECS: 211-743-7 UN: 2929 ADR: 6.1 (3),II |
Specification Test Results
Solubility | (20 °C) (decomposition) |
Molar Mass | 126.2 g/mol |
Boiling Point | 145 - 148 °C (1013 hPa) |
Flash Point | 33 °C |
Refractive Index | 1.427 (20 °C, 589 nm) |
Density | 0.81 g/cm3 (20 °C) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...