4-Benzylphenol 98%

Supplier: Thermo Fisher Scientific
Warning

121130010 121130050
121130010.EA 42.8 EUR
121130010. 121130050.
4-Benzylphenol 98%
4-Benzylphenol
Formula: C₁₃H₁₂O
Boiling Pt: 198…200°C (10mmHg)
Storage Temperature: Ambient
MDL Number: MFCD00002384
CAS Number: 101-53-1
EINECS: 202-950-3

Order Now

Specification Test Results

Appearance white to beige to brown crystalline powder or crystals
Infrared spectrum Conforms
GC ≥97.5 %
Melting point 80°C to 85°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR