Polyacrylonitrile, M.W. ∼150000
Supplier: Thermo Fisher Scientific
310670250
310671000
310670250.EA
84.5
EUR
310670250.
310671000.
Polyacrylonitrile, M.W. ∼150000
Polyacrylonitrile
Formula:
(C₃H₃N)n Storage Temperature: Ambient |
MDL Number:
MFCD00084395 CAS Number: 25014-41-9 |
Specification Test Results
Appearance | white to yellow powder or chunks |
Infrared spectrum | Conforms |
Average molecular weight | 150000 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...