N,N-Diethylformamide, extra pure for MOF synthesis

Supplier: Thermo Fisher Scientific
Danger

Synonyms: Formic acid diethylamide, Formdiethylamide

407300250 407301000
407300250.EA 62 EUR
407300250. 407301000.
N,N-Diethylformamide, extra pure for MOF synthesis
N,N-Diethylformamide
Formula: C₅H₁₁NO
MW: 101.15 g/mol
Boiling Pt: 176…177°C (760mmHg)
Density: 0.9
Flash Pt: 60°C
Storage Temperature: Ambient
MDL Number: MFCD00003287
CAS Number: 617-84-5
EINECS: 210-533-2
UN: 1993
ADR: 3 III

Order Now

Specification Test Results

Appearance clear colorless to light yellow liquid
Infrared spectrum Conforms
GC ≥99 %
Refractive index 1.4330 to 1.4350 (20°C, 589 nm)
Water ≤0.1 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR