Panobinostat

Supplier: Thermo Fisher Scientific
Danger

467000010 467002500
467000010.EA 623 EUR
467000010. 467002500.
Panobinostat
Panobinostat
Formula: C₂₁H₂₃N₃O₂
Storage Temperature: Refrigerator
MDL Number: MFCD09833242
CAS Number: 404950-80-7
UN: 2811
ADR: 6.1,III

Order Now

Specification Test Results

Appearance almost white to yellow to light brown powder
Infrared spectrum Conforms
HPLC ≥97.5 %
Loss on drying ≤2 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR