Climbazole
Supplier: Thermo Fisher Scientific
|
Warning
|
Synonyms:
1-(4-Chlorophenoxy)-1-(imidazol-1-yl)-3,3-dimethyl-2-butanone
468420050
468420250
468421000
468420050.EA
44.5
EUR
468420050.
468420250.
468421000.
Climbazole
Climbazole
Formula:
C₁₅H₁₇ClN₂O₂ Boiling Pt: 260°C Storage Temperature: Ambient |
MDL Number:
MFCD00055505 CAS Number: 38083-17-9 EINECS: 253-775-4 UN: 3077 ADR: 9,III |
Specification Test Results
Appearance | white to light yellow to light beige crystalline powder or crystals |
Infrared spectrum | Conforms |
HPLC | ≥97.5 % |
Melting point | 94°C to 100°C |
Water | ≤1 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...