Hydroxyzine dihydrochloride

Supplier: Thermo Fisher Scientific
Warning

468440010 468440050
468440010.EA 49.8 EUR
468440010. 468440050.
Hydroxyzine dihydrochloride
Hydroxyzine dihydrochloride
Formula: C₂₁H₂₇ClN₂O₂·2HCl
Storage Temperature: Ambient
MDL Number: MFCD00058200
CAS Number: 2192-20-3
EINECS: 218-586-3
Merck Index: 13,04875

Order Now

Specification Test Results

Appearance white powder
Infrared spectrum Conforms
HPLC ≥97.5 %
Titration with HClO4 97.5 to 102.5 % (dry substance)
Loss on drying ≤5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR