Rebamipide
Supplier: Thermo Fisher Scientific
|
Danger
|
Synonyms:
2-(4-Chlorobenzamido)-3-[2(1H)-quinolinon-4-yl]propionic Acid
468590010
468590050
468590250
468590010.EA
56.3
EUR
468590010.
468590050.
468590250.
Rebamipide
Rebamipide
Formula:
C₁₉H₁₅ClN₂O₄ MW: 370.79 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD11114396 CAS Number: 90098-04-7 UN: 2811 ADR: 6.1 III Merck Index: 13,08209 |
Specification Test Results
Appearance | white to beige crystalline powder |
Infrared spectrum | Conforms |
HPLC | ≥98.5 % |
Loss on drying | ≤2 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...