Penciclovir
Supplier: Thermo Fisher Scientific
|
Warning
|
Synonyms:
2-Amino-1,9-dihydro-9-[4-hydroxy-3-(hydroxymethyl)butyl]-6H-purin-6-one, 2-Amino-9-[4-hydroxy-3-(hydroxymethyl)butyl]-1H-purin-6-one, 9-[4-Hydroxy-3-(hydroxymethyl)butyl]guanine, BRL 39123, 2-Amino-9-(4-hydroxy-3-(hydroxymethyl)butyl)-1H-purin-6(9H)-one
468730010
468732500
468730010.EA
373
EUR
468730010.
468732500.
Penciclovir
Penciclovir
Formula:
C₁₀H₁₅N₅O₃ MW: 253.26 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD15142866 CAS Number: 39809-25-1 Merck Index: 13,07154 |
Specification Test Results
Appearance | white to almost white powder |
Infrared spectrum | Conforms |
HPLC | ≥97.5 % |
Loss on drying | ≤1 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...