Sertaconazole nitrate 98%

Supplier: Thermo Fisher Scientific
Warning

469250010 469252500
469250010.EA 155 EUR
469250010. 469252500.
Sertaconazole nitrate 98%
Sertaconazole nitrate
Formula: C₂₀H₁₅Cl₃N₂OS·HNO₃
Storage Temperature: Ambient
MDL Number: MFCD00918063
CAS Number: 99592-39-9
Merck Index: 13,08539

Order Now

Specification Test Results

Appearance white to beige powder
Infrared spectrum Conforms
HPLC ≥97.5 %
Melting point 156°C to 165°C
Water ≤1 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR