Propiconazole mixture of isomers 95%
Supplier: Thermo Fisher Scientific
|
Danger
|
470060010
470062500
470060010.EA
188
EUR
470060010.
470062500.
Propiconazole mixture of isomers 95%
Propiconazole
Formula:
C₁₅H₁₇Cl₂N₃O₂ Boiling Pt: 480°C Density: 1,289 Flash Pt: 11°C Storage Temperature: Ambient |
MDL Number:
MFCD00055299 CAS Number: 60207-90-1 EINECS: 262-104-4 UN: 3082 ADR: 9,III Merck Index: 13,00791 |
Specification Test Results
Appearance | colorless to yellow to light brown clear liquid or viscous liquid |
Infrared spectrum | Conforms |
GC | ≥94 % (mixture of isomers) |
Refractive index | 1.5440 to 1.5480 (20°C, 589 nm) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...