1-Methylpyrene 97%

Supplier: Thermo Fisher Scientific
Warning

470140010 470142500
470140010.EA 292 EUR
470140010. 470142500.
1-Methylpyrene 97%
1-Methylpyrene
Formula: C₁₇H₁₂
Storage Temperature: Ambient
MDL Number: MFCD00014329
CAS Number: 2381-21-7
EINECS: 219-178-8

Order Now

Specification Test Results

Appearance white to brown powder or crystals
Infrared spectrum Conforms
GC ≥96 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR