Enalaprilat dihydrate 95%

Supplier: Thermo Fisher Scientific
Warning

470180010 470182500
470180010.EA 793 EUR
470180010. 470182500.
Enalaprilat dihydrate 95%
Enalaprilat dihydrate
Formula: C₁₈H₂₄N₂O₅·2H₂O
Storage Temperature: Refrigerator
MDL Number: MFCD00941393
CAS Number: 84680-54-6

Order Now

Specification Test Results

Appearance white to beige crystalline powder
Infrared spectrum Conforms
HPLC ≥94 %
Water 7 to 11 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR