Mangiferin 97%

Supplier: Thermo Fisher Scientific
Warning

471790010 471790050
471790010.EA 50.9 EUR
471790010. 471790050.
Mangiferin 97%
Mangiferin
Formula: C₁₉H₁₈O₁₁
Storage Temperature: Refrigerator
MDL Number: MFCD00075656
CAS Number: 4773-96-0

Order Now

Specification Test Results

Appearance white to light yellow powder
Infrared spectrum Conforms
HPLC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR