Glabridin 98%

Supplier: Thermo Fisher Scientific
Warning

471850010 471852500
471850010.EA 315 EUR
471850010. 471852500.
Glabridin 98%
Glabridin
Formula: C₂₀H₂₀O₄
Flash Pt: 267°C
Storage Temperature: Ambient
MDL Number: MFCD03427694
CAS Number: 59870-68-7

Order Now

Specification Test Results

Appearance white to yellow to light brown powder
Infrared spectrum Conforms
HPLC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR